
N,N'-Bis(salicylidene)-1,2-propanediamine ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:94-91-7
:
Formula:C₁₇H₁₈N₂O₂
MDL Number:MFCD00020104
:282.34 g/mol
EINECS:202-374-2
:GHS07
Specification Test Results
- Appearance:Light orange to Yellow to Green powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:45.0 to 51.0 deg-C
- Solubility in Toluene:almost transparency
Specification Test Results
Frequently Bought Together