Order Entry
Netherlands
ContactUsLinkComponent
2,6-Dimethyl-1-heptene ≥97.0% (by GC)
2,6-Dimethyl-1-heptene ≥97.0% (by GC)
  TCIAD1259-1ML
 :  TCI
 :  
undefined
2,6-Dimethyl-1-heptene ≥97.0% (by GC)
  TCIAD1259-1ML
 :  TCI
 :  

 

  • Pk:
    1 mL
  • Seq:
    2070

 

 

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 97.0 %

 

 

 :
 :126.24 g/mol
 :142°C
 :30°C
 :3295
 :3,III
 :GHS02
 :GHS08
 :3074-78-0
 :C₉H₁₈
 :MFCD00048377