

2,4-Dimethyl-1-heptene ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:19549-87-2
:
Formula:C₉H₁₈
MDL Number:MFCD00059250
:126.24 g/mol
Boiling Pt:134°C
UN:3295
ADR:3,II
:GHS02
:GHS08
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together