
1,3-Dibenzylidene-2-cyclohexanone ≥98.0% (by HPLC)
Catalog # TCIAD1194-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:Light yellow to Yellow to Orange powder to crystal
- Purity (HPLC):min. 98.0 area%
- Melting point:116.0 to 119.0 deg-C
Specification Test Results
About this item
:
:274.36 g/mol
EINECS:212-981-4
UN:3077
ADR:9,III
:GHS09
CAS Number:897-78-9
Formula:C₂₀H₁₈O
MDL Number:MFCD00019516