
2,5-Dichlorophenylthioglycolic acid ≥98.0% (by GC, titration analysis)
TCIAD1146-25G
: TCI
:
- Pk:25 g
- Seq:3308
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:129.0 to 133.0 deg-C
:
:237.09 g/mol
:228-468-3
:GHS07
:6274-27-7
:C₈H₆Cl₂O₂S
:MFCD00044041