Order Entry
Netherlands
ContactUsLinkComponent
N-(2,4-Dinitrophenyl)-DL-threonine ≥99.0% (by titrimetric analysis)
Flammable
N-(2,4-Dinitrophenyl)-DL-threonine ≥99.0% (by titrimetric analysis)
Catalog # TCIAD1039-100MG
Supplier:  TCI
CAS Number:  
Flammable
N-(2,4-Dinitrophenyl)-DL-threonine ≥99.0% (by titrimetric analysis)
Catalog # TCIAD1039-100MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    Light yellow to Yellow to Orange powder to crystal
  • Purity (Neutralization titration):
    min. 99.0 %
  • Melting point:
    153.0 to 156.0 deg-C

Specification Test Results

About this item

 :
 :285.21 g/mol
UN:1325
ADR:4.1,II
 :GHS02
CAS Number:14401-07-1
Formula:C₁₀H₁₁N₃O₇
MDL Number:MFCD00070530