
N-(2,4-Dinitrophenyl)-L-serine ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAD1037-100MG
Supplier: TCI
CAS Number:
Specifications
- Pk:100 mg
- Seq:2440
Specifications
Specification Test Results
- Appearance:Light yellow to Yellow to Orange powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Melting point:177.0 to 181.0 deg-C
- Specific rotation [a]20/D:-22.0 to -24.0 deg (C=0.5, AcOH)
Specification Test Results
About this item
:
:271.19 g/mol
EINECS:216-745-1
UN:1325
ADR:4.1,II
:GHS02
CAS Number:1655-64-7
Formula:C₉H₉N₃O₇
MDL Number:MFCD00038098