Order Entry
Netherlands
ContactUsLinkComponent
 
 
N-(2,4-Dinitrophenyl)-L-proline ≥98.0% (by HPLC, titration analysis)
 :  TCI
 
 
N-(2,4-Dinitrophenyl)-L-proline ≥98.0% (by HPLC, titration analysis)
 :  TCI

 

  • Appearance:
    Light yellow to Yellow to Orange powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Neutralization titration):
    min. 98.0 %
  • Melting point:
    138.0 to 141.0 deg-C
  • Specific rotation [a]20/D:
    -675.0 to -690.0 deg (C=0.5, AcOH)

 

Frequently Bought Together