
N-(2,4-Dinitrophenyl)-L-alanine ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAD1031-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:Light yellow to Yellow to Orange powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Melting point:177.0 to 181.0 deg-C
- Specific rotation [a]20/D:+140.0 to +147.0 deg (C=0.5, 1mol/L NaOH sol.)
Specification Test Results
About this item
:
:255.19 g/mol
EINECS:216-739-9
UN:1325
ADR:4.1,II
:GHS02
CAS Number:1655-52-3
Formula:C₉H₉N₃O₆
MDL Number:MFCD00038106