Order Entry
Netherlands
ContactUsLinkComponent
3,9-Divinyl-2,4,8,10-tetraoxaspiro[5.5]undecane ≥98.0% (by GC)
3,9-Divinyl-2,4,8,10-tetraoxaspiro[5.5]undecane ≥98.0% (by GC)
Supplier:  TCI
Certificates
3,9-Divinyl-2,4,8,10-tetraoxaspiro[5.5]undecane ≥98.0% (by GC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    42.0 to 46.0 deg-C
  • Solubility in Methanol:
    almost transparency

Specification Test Results

Related Information

Frequently Bought Together