Order Entry
Netherlands
ContactUsLinkComponent
1,4-Diphenyl-3-thiosemicarbazide ≥99.0% (by titrimetric analysis)
Irritant
1,4-Diphenyl-3-thiosemicarbazide ≥99.0% (by titrimetric analysis)
Supplier:  TCI
Certificates
Irritant
1,4-Diphenyl-3-thiosemicarbazide ≥99.0% (by titrimetric analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Light gray to Light yellow powder to crystal
  • Purity (Neutralization titration):
    min. 99.0 %
  • Melting point:
    171.0 to 181.0 deg-C
  • Solubility in hot Methanol:
    almost transparency

Specification Test Results

Frequently Bought Together