


tert-Dodecyl mercaptan (mixture of isomers) ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:25103-58-6
:
Formula:C₁₂H₂₆S
MDL Number:MFCD00043233
:202.40 g/mol
EINECS:246-619-1
UN:3082
ADR:9,III
:GHS07
:GHS08
:GHS09
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (Neutralization titration):min. 98.0 %
Specification Test Results
Frequently Bought Together