
Disodium-4,4'-dinitrostilbene-2,2'-disulphonate ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:3709-43-1
:
Formula:C₁₄H₁₀N₂O₁₀S₂
MDL Number:MFCD00024929
:474.32 g/mol
EINECS:223-051-2
:GHS07
Specification Test Results
- Appearance:Light yellow to Brown powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Ion exchange titration):min. 98.0 % (calcd.on anh.substance)
- Solubility in Water:almost transparency
- Water:max. 5.0 %
Specification Test Results
Frequently Bought Together