
2-(2,4-Dinitrophenylthio)benzothiazole ≥98.0% (by total nitrogen basis)
Supplier: TCI
Certificates
About this item
CAS Number:17586-89-9
:
Formula:C₁₃H₇N₃O₄S₂
MDL Number:MFCD00059704
:333.34 g/mol
UN:1325
ADR:4.1,III
:GHS02
Specification Test Results
- Appearance:Light yellow to Brown powder to crystal
- Purity (with Total Nitrogen):min. 98.0 %
- Melting point:159.0 to 163.0 deg-C
- Solubility in hot Toluene:faint turbidity
Specification Test Results
Frequently Bought Together