Order Entry
Netherlands
ContactUsLinkComponent
N,N'-Di-2-naphthyl-1,4-phenylenediamine ≥98.0% (by HPLC)
Irritant
N,N'-Di-2-naphthyl-1,4-phenylenediamine ≥98.0% (by HPLC)
Supplier:  TCI
Certificates
Irritant
N,N'-Di-2-naphthyl-1,4-phenylenediamine ≥98.0% (by HPLC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Gray to Brown powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (with Total Nitrogen):
    min. 96.0 %

Specification Test Results

Related Information

Frequently Bought Together