Order Entry
Netherlands
ContactUsLinkComponent
2,3-Dimethylaniline hydrochloride ≥98.0% (by HPLC, total nitrogen)
2,3-Dimethylaniline hydrochloride ≥98.0% (by HPLC, total nitrogen)
Catalog # TCIAD0673-25G
Supplier:  TCI
CAS Number:  
2,3-Dimethylaniline hydrochloride ≥98.0% (by HPLC, total nitrogen)
Catalog # TCIAD0673-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (with Total Nitrogen):
    min. 98.0 %
  • Solubility in Water:
    very faint turbidity

Specification Test Results

About this item

 :
 :157.64 g/mol
CAS Number:5417-45-8
Formula:C₈H₁₁N
MDL Number:MFCD00054332