
Trisodium 2-(4-Sulphophenylazo)-1,8-dihydroxynaphthalene-3,6-disulphonate (SPADNS) ≥95.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:23647-14-5
:
Formula:C₁₆H₁₂N₂O₁₁S₃
MDL Number:MFCD00003952
:570.40 g/mol
EINECS:245-803-9
:GHS07
Specification Test Results
- Appearance:Orange to Brown to Dark red powder to crystal
- Purity (HPLC):min. 95.0 area%
- Water:3.0 to 14.0 %
- Sensitiveness:to pass test
Specification Test Results
Frequently Bought Together