Order Entry
Netherlands
ContactUsLinkComponent
Sodium-6,7-dihydroxynaphthalene-2-sulphonate ≥98.0% (by HPLC, titration analysis)
Irritant
Sodium-6,7-dihydroxynaphthalene-2-sulphonate ≥98.0% (by HPLC, titration analysis)
Supplier:  TCI
Certificates
Irritant
Sodium-6,7-dihydroxynaphthalene-2-sulphonate ≥98.0% (by HPLC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Light yellow to Light orange powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Ion exchange titration):
    min. 98.0 % (calcd.on anh.substance)
  • Solubility in Water:
    almost transparency
  • Water:
    max. 5.0 %

Specification Test Results

Related Information

Frequently Bought Together