Order Entry
Netherlands
ContactUsLinkComponent
 
 
 
1,8-Dihydroxy-9,10-anthraquinone ≥98.0% (by GC, titration analysis)
  TCIAD0563-25G
 :  TCI
 :  
 
 
1,8-Dihydroxy-9,10-anthraquinone ≥98.0% (by GC, titration analysis)
  TCIAD0563-25G
 :  TCI
 :  

 

  • Pk:
    25 g
  • Seq:
    3308

 

 

  • Appearance:
    Light yellow to Brown powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Purity (Neutralization titration):
    min. 98.0 %
  • Melting point:
    191.0 to 196.0 deg-C
  • Solubility in hot Acetic acid:
    very faint turbidity

 

 

 :
 :240.22 g/mol
 :14,02815
 :204-173-5
 :GHS07
 :GHS08
 :117-10-2
 :C₁₄H₈O₄
 :MFCD00001211