Order Entry
Netherlands
ContactUsLinkComponent
4,4'-Didodecyloxyazoxybenzene ≥98.0% (by total nitrogen basis)
4,4'-Didodecyloxyazoxybenzene ≥98.0% (by total nitrogen basis)
Catalog # TCIAD0553-5G
Supplier:  TCI
CAS Number:  
4,4'-Didodecyloxyazoxybenzene ≥98.0% (by total nitrogen basis)
Catalog # TCIAD0553-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    Light yellow to Yellow to Orange powder to crystal
  • Purity (with Total Nitrogen):
    min. 98.0 %
  • Mesomorphic range:
    80.0 to 126.0 deg-C

Specification Test Results

About this item

 :
 :566.87 g/mol
CAS Number:2312-14-3
Formula:C₃₆H₅₈N₂O₃
MDL Number:MFCD00059283