Order Entry
Netherlands
ContactUsLinkComponent
 
 
 
 
Diethylstilbestrol cis- and trans-mixture ≥98.0% (by GC)
 :  TCI
 
 
 
 
Diethylstilbestrol cis- and trans-mixture ≥98.0% (by GC)
 :  TCI

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    169.0 to 173.0 deg-C
  • Solubility in Methanol:
    almost transparency

 

Frequently Bought Together