
N,N-Diethyl-1,4-phenylenediamine dihydrochloride ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:16713-15-8
:
Formula:C₁₀H₁₆N₂
MDL Number:MFCD00060227
:237.17 g/mol
EINECS:240-760-2
:GHS07
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (Neutralization titration):min. 98.0 %
- Solubility in Water:almost transparency
Specification Test Results
Frequently Bought Together