
2,4-Diamino-6-diethylamino-1,3,5-triazine ≥98.0% (by titrimetric analysis)
Catalog # TCIAD0512-25G
Supplier: TCI
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:168.0 to 171.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
About this item
:
:182.23 g/mol
:GHS07
CAS Number:2073-31-6
Formula:C₇H₁₄N₆
MDL Number:MFCD00059765