
DL-2,3-Diaminopropionic acid hydrochloride ≥98.0% (by total nitrogen basis)
: TCI
:54897-59-5
:
:C₃H₈N₂O₂
:MFCD00012884
:140.57 g/mol
:259-387-1
:GHS07
- Appearance:White to Light yellow powder to crystal
- Purity (with Total Nitrogen):min. 98.0 %
- Solubility in Water:almost transparency
Frequently Bought Together