Order Entry
Netherlands
ContactUsLinkComponent
 
Sodium-2,5-dichlorosulphanilate ≥95.0% (by HPLC, titration analysis)
  TCIAD0416-25G
 :  TCI
 :  
Sodium-2,5-dichlorosulphanilate ≥95.0% (by HPLC, titration analysis)
  TCIAD0416-25G
 :  TCI
 :  

 

  • Pk:
    25 g
  • Seq:
    3308

 

 

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (HPLC):
    min. 95.0 area%
  • Purity (Ion exchange titration):
    min. 95.0 %
  • Water:
    max. 7.0 %

 

 

 :
 :264.05 g/mol
 :255-302-7
 :41295-98-1
 :C₆H₅Cl₂NO₃S
 :MFCD00068356