

2,3-Dichloropropionic acid ≥98.0% (by GC, titration analysis)
: TCI
:565-64-0
:
:C₃H₄Cl₂O₂
:MFCD00021695
:142.96 g/mol
:210°C
:209-285-8
:2923
:8,II
:GHS05
:GHS06
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:51.0 to 55.0 deg-C
- Solubility in Water:almost transparency
Frequently Bought Together