Order Entry
Netherlands
ContactUsLinkComponent
 
N,N'-Dibenzoylhydrazine ≥95.0% (by titrimetric analysis)
 :  TCI
 
N,N'-Dibenzoylhydrazine ≥95.0% (by titrimetric analysis)
 :  TCI

 

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (Bromination Method):
    min. 95.0 %

 

Frequently Bought Together