
1,1'-Iminodianthraquinone ≥98.0% (by total nitrogen basis) for determination of boron
Catalog # TCIAD0133-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:Orange to Brown to Dark red powder to crystal
- Purity (with Total Nitrogen):min. 98.0 %
Specification Test Results
About this item
:
:429.43 g/mol
Merck Index:14,00690
EINECS:201-405-7
:GHS07
CAS Number:82-22-4
Formula:C₂₈H₁₅NO₄
MDL Number:MFCD00019132