
2,5-Diaminotoluene dihydrochloride ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAD0126-25G
Supplier: TCI
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Gray to Red powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Argentmetric Titration):min. 98.0 %
Specification Test Results
About this item
:
ADR:6.1,III
:195.09 g/mol
UN:2811
:GHS06
CAS Number:615-45-2
Formula:C₇H₁₀N₂
MDL Number:MFCD00035512