Order Entry
Netherlands
ContactUsLinkComponent
2-Cyclohexylpropan-2-yl methacrylate ≥98.0% (by GC) stabilised
2-Cyclohexylpropan-2-yl methacrylate ≥98.0% (by GC) stabilised
  TCIAC3642-1G
 :  TCI
 :  
2-Cyclohexylpropan-2-yl methacrylate ≥98.0% (by GC) stabilised
  TCIAC3642-1G
 :  TCI
 :  

 

  • Pk:
    1 g
  • Seq:
    3047

 

 

  • Appearance:
    Colorless to Light yellow clear liquid
  • Purity (GC):
    min. 98.0 %

 

 

 :
 :210.32 g/mol
 :Stabilised with phenothiazine
 :186585-56-8
 :C₁₃H₂₂O₂