


2-[(Chloromethoxy)methyl]naphthalene ∼2 mol/L in toluene
TCIAC3639-5ML
: TCI
:
- Pk:5 mL
- Seq:2210
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Concentration (HNMR):1.9 to 2.5 mol/L
:
:206.67 g/mol
:Refrigerator
:1294
:3,II
:GHS02
:GHS07
:GHS08
:914300-10-0
:C₁₂H₁₁ClO
:MFCD30741917