
2-Chloro-3-fluorophenylboronic acid (contains varying amounts of Anhydride)
TCIAC3563-1G
: TCI
:
- Pk:1 g/mol
- Seq:9220
- Appearance:White to Light yellow powder to crystal
- Purity (Neutralization titration):97.0 to 110.0 %
:
:174.37 g/mol
:GHS07
:871329-52-1
:C₆H₅BClFO₂
:MFCD07363741