Order Entry
Netherlands
ContactUsLinkComponent
4-Cyano-4-[[(dodecylthio)carbonothioyl]thio]pentanoic acid ≥97.0% (by HPLC, titration analysis)
Irritant
4-Cyano-4-[[(dodecylthio)carbonothioyl]thio]pentanoic acid ≥97.0% (by HPLC, titration analysis)
Supplier:  TCI
Certificates
Irritant
4-Cyano-4-[[(dodecylthio)carbonothioyl]thio]pentanoic acid ≥97.0% (by HPLC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Light yellow to Light orange powder to crystal
  • Purity (HPLC):
    min. 97.0 area%
  • Purity (Neutralization titration):
    min. 97.0 %
  • Melting point:
    61.0 to 65.0 deg-C

Specification Test Results

Frequently Bought Together