2-Chloro-4,6-bis(9,9-dimethyl-9H-fluoren-2-yl)-1,3,5-triazine ≥98.0% (by total nitrogen basis)
Supplier: TCI
Certificates
About this item
CAS Number:1459162-69-6
:
Formula:C₃₃H₂₆ClN₃
MDL Number:MFCD30471507
:500.04 g/mol
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC):min. 97.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
Specification Test Results
Frequently Bought Together