
3-(4'-Chlorophenyl)propanol ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:6282-88-8
:
Formula:C₉H₁₁ClO
MDL Number:MFCD00831102
:170.64 g/mol
Boiling Pt:114°C
:GHS07
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together