H-D-Lys(Z)-OH ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:34404-32-5
:
Formula:C₁₄H₂₀N₂O₄
MDL Number:MFCD00063120
:280.324 g/mol
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
- Specific rotation [a]20/D:-15.0 to -17.0 deg (C=1.6,2mol/L HCl)
Specification Test Results
Frequently Bought Together