
4-(4-Chlorophenyl)piperidine ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:26905-02-2
:
Formula:C₁₁H₁₄ClN
MDL Number:MFCD03077010
:195.69 g/mol
Boiling Pt:160°C
:GHS07
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (GC):min. 97.0 %
- Melting point:204.0 to 208.0 deg-C
Specification Test Results
Frequently Bought Together