
2'-Chloro-5'-(trifluoromethyl)acetophenone ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:71648-45-8
:
Formula:C₉H₆ClF₃O
MDL Number:MFCD00045092
:222.59 g/mol
Flash Pt:91°C
:GHS07
Specification Test Results
- Appearance:Colorless to Light orange to Yellow clear liquid
- Purity (GC):min. 97.0 %
Specification Test Results
Frequently Bought Together