Order Entry
Netherlands
ContactUsLinkComponent
Chloro[(1,2,3,4,5,6-eta)-2,2'',4,4'',6,6''-hexamethyl[1,1':3',1''-terphenyl]-2'-thiolato-kappaS][tris(4-fluorophenyl)phosphine-kappaP]ruthenium(II)
Chloro[(1,2,3,4,5,6-eta)-2,2'',4,4'',6,6''-hexamethyl[1,1':3',1''-terphenyl]-2'-thiolato-kappaS][tris(4-fluorophenyl)phosphine-kappaP]ruthenium(II)
Catalog # TCIAC3328-100MG
Supplier:  TCI
CAS Number:  
undefined
Chloro[(1,2,3,4,5,6-eta)-2,2'',4,4'',6,6''-hexamethyl[1,1':3',1''-terphenyl]-2'-thiolato-kappaS][tris(4-fluorophenyl)phosphine-kappaP]ruthenium(II)
Catalog # TCIAC3328-100MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    Light yellow to Amber to Dark green powder to crystal
  • Elemental analysis (Carbon):
    60.50 to 65.50 %
  • Elemental analysis (Hydrogen):
    4.40 to 5.00 %

Specification Test Results

About this item

 :
 :798.31 g/mol
CAS Number:1420299-84-8
Formula:C₄₂H₃₇ClF₃PRuS