Chloro[(1,2,3,4,5,6-eta)-2,2'',4,4'',6,6''-hexamethyl[1,1':3',1''-terphenyl]-2'-thiolato-kappaS][triisopropylphosphine-kappaP]ruthenium(II)
Catalog # TCIAC3327-100MG
Supplier: TCI
CAS Number:
Specifications
- Pk:100 mg
- Seq:2440
Specifications
Specification Test Results
- Appearance:Light yellow to Amber to Dark green powder to crystal
- Elemental analysis (Carbon):59.00 to 64.00 %
- Elemental analysis (Hydrogen):6.80 to 7.50 %
Specification Test Results
About this item
:
:642.28 g/mol
CAS Number:1621182-04-4
Formula:C₃₃H₄₆ClPRuS