
3-Chloro-5-cyanophenylboronic acid (contains varying amounts of Anhydride)
Catalog # TCIAC3266-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization titration):97.0 to 112.0 %
Specification Test Results
About this item
:
:181.38 g/mol
:GHS07
CAS Number:915763-60-9
Formula:C₇H₅BClNO₂
MDL Number:MFCD09263609