
Cesium Pivalate ≥97.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:20442-70-0
:
Formula:C₅H₉CsO₂
MDL Number:MFCD10567040
:234.03 g/mol
Melting Pt:348°C
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Nonaqueous Titration):min. 97.0 %
Specification Test Results
Frequently Bought Together