
2,4-Difluoro-3-cyanophenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI
Certificates
About this item
CAS Number:871940-31-7
:
Formula:C₇H₄BF₂NO₂
MDL Number:MFCD12026699
Melting Pt:134°C
:182.92 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization titration):97.0 to 111.0 %
Specification Test Results
Frequently Bought Together