
5-Chloro-2-fluorobenzylamine ≥97.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:261723-26-6
:
Formula:C₇H₇ClFN
MDL Number:MFCD01631440
:159.59 g/mol
UN:2735
ADR:8,III
:GHS05
Specification Test Results
- Appearance:Light orange to Yellow to Green clear liquid
- Purity (GC):min. 97.0 %
- Purity (Neutralization titration):min. 97.0 %
Specification Test Results
Frequently Bought Together