Fmoc-Lys(Z)-OH ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:86060-82-4
:
Formula:C₂₉H₃₀N₂O₆
MDL Number:MFCD00065662
:502.567 g/mol
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Melting point:115.0 to 119.0 deg-C
- Specific rotation [a]20/D:-9.0 to -11.0 deg (C=2,DMF)
Specification Test Results
Frequently Bought Together