Z-Pyr-OH ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:32159-21-0
:
Formula:C₁₃H₁₃NO₅
MDL Number:MFCD00037352
:263.249 g/mol
Storage Temperature:Freezer
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Melting point:139.0 to 143.0 deg-C
- Specific rotation [a]20/D:-34.0 to -37.0 deg (C=1,MeOH)
Specification Test Results
Frequently Bought Together