6-Chloroimidazo[1,2-b]pyridazine ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:6775-78-6
:
Formula:C₆H₄ClN₃
MDL Number:MFCD07778345
:153.5691 g/mol
Specification Test Results
- Appearance:White to Gray to Brown powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:114.0 to 118.0 deg-C
Specification Test Results
Frequently Bought Together