
(5-Carboxypentyl)triphenylphosphonium bromide ≥96.0% (by titrimetric analysis)
Catalog # TCIAC3113-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC):min. 95.0 area%
- Purity (Neutralization titration):min. 96.0 %
- Melting point:198.0 to 202.0 deg-C
Specification Test Results
About this item
:
:457.36 g/mol
:GHS07
CAS Number:50889-29-7
Formula:C₂₄H₂₆BrO₂P
MDL Number:MFCD00055556