
(3-Chloropropyl)diethoxy(methyl)silane ≥98.0% (by GC)
: TCI
:13501-76-3
:
:C₈H₁₉ClO₂Si
:MFCD05663910
:109°C
:210.77 g/mol
:236-828-6
:GHS07
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
Frequently Bought Together