Order Entry
Netherlands
ContactUsLinkComponent
2-(3-Chloropropyl)-1,3-dioxolane ≥97.0% (by GC)
2-(3-Chloropropyl)-1,3-dioxolane ≥97.0% (by GC)
Catalog # TCIAC3033-5G
Supplier:  TCI
CAS Number:  
2-(3-Chloropropyl)-1,3-dioxolane ≥97.0% (by GC)
Catalog # TCIAC3033-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    Colorless to Brown clear liquid
  • Purity (GC):
    min. 97.0 %

Specification Test Results

About this item

 :
 :150.6 g/mol
Boiling Pt:53°C
Flash Pt:79°C
CAS Number:16686-11-6
Formula:C₆H₁₁ClO₂
MDL Number:MFCD00043135